$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
BRUSH GREEN 285X130MM
85013-200
:
FBK MEXICO, S.A. DE C.V. MX
BRUSH GREEN 285X130MM
85013-200
:
FBK MEXICO, S.A. DE C.V. MX
:
20154-5
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH