$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
CEPILLO PEQUE#O DE MANO CON MANGO, VERDE
85013-182
:
FBK MEXICO, S.A. DE C.V. MX
CEPILLO PEQUE#O DE MANO CON MANGO, VERDE
85013-182
:
FBK MEXICO, S.A. DE C.V. MX
:
10463-5
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Universal Pipette Tips
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International