Order Entry
Mexico
Orders LinkContactUsLinkComponent
Float Racks for Microtubes, Globe Scientific

Float Racks for Microtubes, Globe Scientific

65 mm (2.5 dia), White, 8
Catalog # 76169-974
Supplier #:  456398
Float Racks for Microtubes, Globe Scientific
Catalog # 76169-974
Float Racks for Microtubes, Globe Scientific
Float Racks for Microtubes, Globe Scientific

65 mm (2.5 dia), White, 8

Price
$652.07
Log in for availability
Paquete de 4
Product Details & Documents

Autoclavable polypropylene construction for strength, buoyancy, and temperature resistance.

  • Racks float with full load of filled tubes
  • Molded handle in center of rack for comfort and support during transport
  • Legs allow rack to stand upright for use on benchtop
  • Use in ice, water baths, or on the benchtop
  • 65 mm round rack fits most 400 mL beakers

Commonly found in cloning, microbiology, and immunology labs.

Specifications
  • No. of Holes:
    8
  • Color:
    White
  • Description:
    65 mm (2.5 dia)