Float Racks for Microtubes, Globe Scientific
95 x 95 x 80 mm (3.75 x 3.75 x 3"), White, 16
76169-972
: 456395
Autoclavable polypropylene construction for strength, buoyancy, and temperature resistance.
- Racks float with full load of filled tubes
- Molded handle in center of rack for comfort and support during transport
- Legs allow rack to stand upright for use on benchtop
- Use in ice, water baths, or on the benchtop
- 65 mm round rack fits most 400 mL beakers
Commonly found in cloning, microbiology, and immunology labs.
- No. of Holes:16
- Color:White
- Description:95 x 95 x 80 mm (3.75 x 3.75 x 3")
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



