Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward’s® Adjustable Volume Micropipettes
1000 μL Micropipette, 100 - 1000 µl, 5.0 µl
  470231-602
 :  7010301014
Ward’s® Adjustable Volume Micropipettes
  470231-602
Ward’s® Adjustable Volume Micropipettes
Ward’s® Adjustable Volume Micropipettes
1000 μL Micropipette, 100 - 1000 µl, 5.0 µl
$1,606.62
Each (1UN)
The Wards® Micropipette line is fully autoclavable, helping to prevent cross contamination. The ergonomic design is ambidextrous, providing all users with an excellent operating experience.
  • Fully autoclavable
  • Slim, lightweight ergonomic design
  • Compatible with most universal tip brands
  • Tool included for quick and easy recalibration
  • Purchase individually, or as one of our available sets
Personal Pipette Set comes with one of each pipette shown, plus a storage carousel.
Classroom Pipette Set comes with eight of each pipette shown, plus eight storage carousels.
  • Volume:
    100 - 1000 µl
  • Description:
    1000 μL Micropipette
  • Increments:
    5.0 µl