Boreal C-SCOPE Compound Microscopes
Product Details & Documents
A robust microscope that provides crisp and clear visual and digital results!
- Infinity optics
- Plan lead-free objectives, 4, 10, 40 and 100X oil immersion
- Rackless mechanical stage
- Custom camera packages available for Trinocular model
The new Boreal C-SCOPE models are setting a new standard in microscope performance through improvements both in optical and mechanical features. Designed to be used in educational life science, medical and a variety of biological applications, these new microscopes are built around Boreal's complete understanding of the daily demands placed on a standard educational microscope.
Student proof features together with Infinity Plan optics provide fully corrected, repeatable, quality images.
Ordering information: The camera packages for microscope 470231-464 are sold separately and include everything needed to connect to the microscope.
Documents
Specifications
- Description:Trinocular
- Cat. No.:470231-464
- Objectives:4×, 10×, 40×, 100×
- Illumination:3W LED
- Stage:Rackless mechanical