Order Entry
Mexico
Orders LinkContactUsLinkComponent
Boreal C-SCOPE Compound Microscopes
Boreal C-SCOPE Compound Microscopes
Binocular
  470231-462
Supplier #:  1100100403313
Boreal C-SCOPE Compound Microscopes
  470231-462
Boreal C-SCOPE Compound Microscopes
Boreal C-SCOPE Compound Microscopes
Binocular
$23,932.91
Each (1UN)
A robust microscope that provides crisp and clear visual and digital results!
  • Infinity optics
  • Plan lead-free objectives, 4, 10, 40 and 100X oil immersion
  • Rackless mechanical stage
  • Custom camera packages available for Trinocular model
The new Boreal C-SCOPE models are setting a new standard in microscope performance through improvements both in optical and mechanical features. Designed to be used in educational life science, medical and a variety of biological applications, these new microscopes are built around Boreal's complete understanding of the daily demands placed on a standard educational microscope.
Student proof features together with Infinity Plan optics provide fully corrected, repeatable, quality images.
Ordering information: The camera packages for microscope 470231-464 are sold separately and include everything needed to connect to the microscope.
  • Description:
    Binocular
  • Cat. No.:
    470231-462
  • Objectives:
    4×, 10×, 40×, 100×
  • Illumination:
    3W LED
  • Stage:
    Rackless mechanical