Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
POTASSIUM TELLURITE SOLUTION 10X10 MLF
Catalog # 85008-444
Supplier:
SIGMA ALDRICH QUIMICA,S DE RL MX
Expand image
POTASSIUM TELLURITE SOLUTION 10X10 MLF
Catalog # 85008-444
Supplier:
SIGMA ALDRICH QUIMICA,S DE RL MX
Supplier Number:
17774-10X10MLF
Frequently Bought Together
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International