Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Registrarse
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BRUSH HAND CARVING PES WHITE 210 X 70 MM
Catalog # 85008-430
Supplier:
FBK MEXICO, S.A. DE C.V. MX
Expand image
BRUSH HAND CARVING PES WHITE 210 X 70 MM
Catalog # 85008-430
Supplier:
FBK MEXICO, S.A. DE C.V. MX
Supplier Number:
15063-1
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH