$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
INCUBATOR MICRO.D.SEG. 104L
85008-590
:
Thermo Fisher Scientific
INCUBATOR MICRO.D.SEG. 104L
85008-590
:
Thermo Fisher Scientific
:
51028070
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International