Saltar a Encabezado
Saltar a Contenido principal
Saltar a Pie de página
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SENSOR DE TEMP CUB./PT1000.90
Catalog # 85007-060
Supplier:
IKA WORKS, INC. MX
Expand image
SENSOR DE TEMP CUB./PT1000.90
Catalog # 85007-060
Supplier:
IKA WORKS, INC. MX
Supplier Number:
4480600
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH