$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
BURETA CL B LLAVE LATERAL 10
85004-364
:
BRAND GMBH & CO. MX
BURETA CL B LLAVE LATERAL 10
85004-364
:
BRAND GMBH & CO. MX
:
13563
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Disposable Serological Pipettes
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International