Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BURETA ACCU-RED CERT.LOTE
Catalog # 85004-324
Supplier:
Corning
Expand image
BURETA ACCU-RED CERT.LOTE
Catalog # 85004-324
Supplier:
Corning
Supplier Number:
MX2103C-10
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International