Skip to Header
Skip to Main Content
Skip to Footer
Avanzado
Avanzado
SDS
Certificados
Estructura química
Datos del fabricante
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BOTE 115ML C/TAPA DESLIZANT PP
Catalog # 85004-214
Supplier:
BRAND GMBH & CO. MX
Expand image
BOTE 115ML C/TAPA DESLIZANT PP
Catalog # 85004-214
Supplier:
BRAND GMBH & CO. MX
Supplier Number:
61820
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International