Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FIXED-ANGLE ROTOR FIBERLITE F23-48 X 1.5
Catalog # 85003-638
Supplier:
THERMO FISHER SCIENTIFIC MX
Expand image
FIXED-ANGLE ROTOR FIBERLITE F23-48 X 1.5
Catalog # 85003-638
Supplier:
THERMO FISHER SCIENTIFIC MX
Supplier Number:
MX096-484075
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International