$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
REGIST TEMPERATURE / RH 24HR 8IN
Catalog # 85003-606
Supplier:
GRAINGER,S.A. DE C.V. MX
Expand image
REGIST TEMPERATURE / RH 24HR 8IN
Catalog # 85003-606
Supplier:
GRAINGER,S.A. DE C.V. MX
Supplier Number:
MXB01PRU6
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International