Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MICRONQUICK SUPERIOR CLEANING GN 40 X 38
Catalog # 85003-418
Supplier:
FREUDENBERG PRODUCTOS DEL HO- MX
Expand image
MICRONQUICK SUPERIOR CLEANING GN 40 X 38
Catalog # 85003-418
Supplier:
FREUDENBERG PRODUCTOS DEL HO- MX
Supplier Number:
VIL152108
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International