Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Registrarse
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
O-RING 0.487 INNER-DIAMETERX0.103 EPR 60
Catalog # 85003-386
Supplier:
THERMO FISHER SCIENTIFIC MX
Expand image
O-RING 0.487 INNER-DIAMETERX0.103 EPR 60
Catalog # 85003-386
Supplier:
THERMO FISHER SCIENTIFIC MX
Supplier Number:
MXGSX29
Frequently Bought Together
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH