Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HANDLE SUPER HYGIENIC RED 1700 X 32 MM
85003-194
:
FBK MEXICO, S.A. DE C.V. MX
HANDLE SUPER HYGIENIC RED 1700 X 32 MM
85003-194
:
FBK MEXICO, S.A. DE C.V. MX
:
MX29905-3
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
VWR® Universal Pipette Tips
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International