Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Registrarse
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BROOM SWEEP HARD POLYESTER RED 500X60 MM
Catalog # 85002-766
Supplier:
FBK MEXICO, S.A. DE C.V. MX
Expand image
BROOM SWEEP HARD POLYESTER RED 500X60 MM
Catalog # 85002-766
Supplier:
FBK MEXICO, S.A. DE C.V. MX
Supplier Number:
MX26190-3
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International