$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
BRUSH SOFT POLYESTER YELLOW 290 X 130 MM
85002-618
:
FBK MEXICO, S.A. DE C.V. MX
BRUSH SOFT POLYESTER YELLOW 290 X 130 MM
85002-618
:
FBK MEXICO, S.A. DE C.V. MX
:
MX29105-4
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH