$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
AMMON HYDROXIDE 30% RE 4L
Catalog # 85002-144
Supplier:
Avantor
Expand image
AMMON HYDROXIDE 30% RE 4L
Catalog # 85002-144
Supplier:
Avantor
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International