Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
INCUBATOR MICRO.D.SEG. 104L
Catalog # 85003-010
Supplier:
Thermo Fisher Scientific
Expand image
INCUBATOR MICRO.D.SEG. 104L
Catalog # 85003-010
Supplier:
Thermo Fisher Scientific
Supplier Number:
51028070
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International