$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
COVER CUPSINK RESIN BK 7.44X4.44X0.32IN
85002-674
:
LABCONCO CORPORATION MX
COVER CUPSINK RESIN BK 7.44X4.44X0.32IN
85002-674
:
LABCONCO CORPORATION MX
:
4005201
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Disposable Serological Pipettes
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International