DR-MED® Laparoscopic Training Instruments
Laparoscopic forceps
Catalog # 470231-228
Supplier: DR INSTRUMENTS INC. SE
Supplier #: DR786922
Jump to:
Product Details & Documents
Reasonably priced practice tools.
- The same tools you would use in surgery
- 360 degree rotation
- 19" length
Learn skills using tools with the same features and grips as the surgical instruments you will be using. Securely hold and manipulate tissues with the grasper and perform fine manipulations with the forceps. Practice removing tissue with the scissors, and closing internally with the needle driver.
Caution: For Training Purposes Only. Not for human or animal use.
Specifications
- Description:Laparoscopic forceps