Order Entry
Mexico
Orders LinkContactUsLinkComponent
Eisco® Light Emitting Diodes
Eisco® Light Emitting Diodes
Light Emitting Diode (led), Green, pack 10
Catalog # 470230-384
Supplier #:  PH1019B
Eisco® Light Emitting Diodes
Catalog # 470230-384
Eisco® Light Emitting Diodes
Eisco® Light Emitting Diodes
Light Emitting Diode (led), Green, pack 10
Price
$137.51
Log in for availability
Paquete de 10
Product Details & Documents
Diodes for PCB or panel mount.
  • Red LED 5 mm standard high quality with diffused lens
  • Maximum voltage 3.0 V D.C. PCB or panel mount
  • Pack of 10
Labs Red LED 5 mm standard high quality with diffused lens. Maximum voltage 3.0 V D.C. PCB or panel mount. Pack of 10
Specifications
  • Description:
    Light Emitting Diode (led), Green, pack 10