Order Entry
Mexico
Orders LinkContactUsLinkComponent
Eisco® Electrodes
Nickel electrode, strip, 100×19 mm
  470230-340
 :  PH0927G
Eisco® Electrodes
  470230-340
Eisco® Electrodes
Eisco® Electrodes
Nickel electrode, strip, 100×19 mm
$251.57
Each (1UN)
Individual metal electrodes for electrochemical experiments.
  • Each electrode is stamped with its metal type for easy identification
  • Durable and reusable for multiple lab sessions
Eisco electrodes are an essential tool for students exploring electrochemical reactions and principles. Each electrode strip measures approximately 100×19 mm (4×0.75"), is crafted from high-quality metals and clearly stamped for easy identification.These electrodes are perfect for hands-on learning in topics such as oxidation-reduction reactions, electrode potentials, and galvanic cells.
  • Description:
    Nickel electrode, strip
  • Size:
    100×19 mm