Order Entry
Mexico
Orders LinkContactUsLinkComponent
Friction Rods
Friction Rods
Ebonite Rod
Catalog # 470230-306
Supplier #:  PH0856A
Friction Rods
Catalog # 470230-306
Supplier:  
Friction Rods
Friction Rods
Ebonite Rod
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FRICTION ROD EBONITE 30CM Catalog #470307-036
Product 470307-036 is a replacement for 470230-306 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
For your Electrostatic Needs!
  • Excellent quality
Friction rods are used for producing static charges when rubbed against materials such as wool, flannel, silk, and fur.
Specifications
  • Description:
    Ebonite Rod