Order Entry
Mexico
Orders LinkContactUsLinkComponent
Crystallizing Dish

Crystallizing Dish

40 ml, 50 mm, 30 mm
Catalog # 470229-208
Supplier #:  CH0066D
Crystallizing Dish
Catalog # 470229-208
Crystallizing Dish
Crystallizing Dish

40 ml, 50 mm, 30 mm

Price
$165.77
Log in for availability
Each (1UN)
Product Details & Documents

Ideal for storage and crystallization.

  • Borosilicate glass
  • Flat bottom
  • Easy pour spout

Made from high quality borosilicate glass, these crystallizing dishes are perfect for use in any laboratory. Dishes feature flat bottoms and sides, with an easy pour spout. Available in five sizes.

Specifications
  • Ø:
    50 mm
  • Capacity:
    40 ml
  • Height:
    30 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.