Crystallizing Dish
1000 ml, 150 mm, 75 mm
Catalog # 470229-204
Supplier: EISCO SCIENTIFIC LLC
Supplier #: CH0066B
Jump to:
Product Details & Documents
Ideal for storage and crystallization.
- Borosilicate glass
- Flat bottom
- Easy pour spout
Made from high quality borosilicate glass, these crystallizing dishes are perfect for use in any laboratory. Dishes feature flat bottoms and sides, with an easy pour spout. Available in five sizes.
Specifications
- Ø:150 mm
- Capacity:1000 ml
- Height:75 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



