Order Entry
Mexico
Orders LinkContactUsLinkComponent
Crystallizing Dish

Crystallizing Dish

300 ml, 100 mm, 50 mm
Catalog # 470229-202
Supplier #:  CH0066A
Crystallizing Dish
Catalog # 470229-202
Supplier:  
Crystallizing Dish
Crystallizing Dish

300 ml, 100 mm, 50 mm

Discontinued
Product Details & Documents

Ideal for storage and crystallization.

  • Borosilicate glass
  • Flat bottom
  • Easy pour spout

Made from high quality borosilicate glass, these crystallizing dishes are perfect for use in any laboratory. Dishes feature flat bottoms and sides, with an easy pour spout. Available in five sizes.

Specifications
  • Ø:
    100 mm
  • Capacity:
    300 ml
  • Height:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.