Order Entry
Mexico
Orders LinkContactUsLinkComponent
Volumetric Flasks

Volumetric Flasks

1000 ml, 22, ±0.300 ml
Catalog # 470228-960
Supplier #:  CH0441G
Volumetric Flasks
Catalog # 470228-960
Volumetric Flasks
Volumetric Flasks

1000 ml, 22, ±0.300 ml

Price
$608.70
Log in for availability
Each (1UN)
Product Details & Documents

Class A Flasks.

  • Blue graduations
  • Glass stopper

Perfect for everyday use in the laboratory, these borosilicate glass flasks are marked with blue graduations for easy reading and fitted with a solid glass stopper. In compliance with ASTM-E288 standard.

Specifications
  • Capacity:
    1000 ml
  • Stopper No.:
    22
  • Tolerance:
    ±0.300 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.