Order Entry
Mexico
Orders LinkContactUsLinkComponent
Volumetric Flasks

Volumetric Flasks

500 ml, 19, ±0.200 ml
Catalog # 470228-958
Supplier #:  CH0441F
Volumetric Flasks
Catalog # 470228-958
Volumetric Flasks
Volumetric Flasks

500 ml, 19, ±0.200 ml

Price
$471.16
Log in for availability
Each (1UN)
Product Details & Documents

Class A Flasks.

  • Blue graduations
  • Glass stopper

Perfect for everyday use in the laboratory, these borosilicate glass flasks are marked with blue graduations for easy reading and fitted with a solid glass stopper. In compliance with ASTM-E288 standard.

Specifications
  • Capacity:
    500 ml
  • Stopper No.:
    19
  • Tolerance:
    ±0.200 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.