Order Entry
Mexico
Orders LinkContactUsLinkComponent
Volumetric Flasks

Volumetric Flasks

50 ml, 13, ±0.050 ml
Catalog # 470228-952
Supplier #:  CH0441B
Volumetric Flasks
Catalog # 470228-952
Volumetric Flasks
Volumetric Flasks

50 ml, 13, ±0.050 ml

Price
$270.76
Log in for availability
Each (1UN)
Product Details & Documents

Class A Flasks.

  • Blue graduations
  • Glass stopper

Perfect for everyday use in the laboratory, these borosilicate glass flasks are marked with blue graduations for easy reading and fitted with a solid glass stopper. In compliance with ASTM-E288 standard.

Specifications
  • Capacity:
    50 ml
  • Stopper No.:
    13
  • Tolerance:
    ±0.050 ml