Vinyl Lab Apron
X-Small Apron, Clear, 15×22"
Catalog # 470228-492
Supplier: EISCO SCIENTIFIC LLC
Supplier #: VNAPXS
Jump to:
Product Details & Documents
Tough vinyl lab aprons are resistant to lab reagents, acids, alkalis, oils, and grease.
- Halter Design
- Aprons Do Not Restrict Movement
- Two Sizes Available
- Resistant to Acids, Alkalis, Oils, and More
- Reinforced Strain Points
- Latex free
The halter design keeps arms free for unrestricted movement in the lab.
The aprons are reinforced at strain points.
Specifications
- Color:Clear
- Description:X-Small Apron
- W×L:15×22"