Order Entry
Mexico
Orders LinkContactUsLinkComponent
ProPette™ Electronic Pipette Controller
ProPette™ Electronic Pipette Controller
Charger
Catalog # 470228-488
Supplier:   MTC BIO, INC.
Supplier #:  P6080-RC
ProPette™ Electronic Pipette Controller
Catalog # 470228-488
Supplier:  MTC BIO, INC.
ProPette™ Electronic Pipette Controller
ProPette™ Electronic Pipette Controller
Charger
Price
$794.72
Log in for availability
Each (1UN)
Product Details & Documents
ProPette™ features an ergonomic body design, with computer-balanced weight distribution and low pressure fingertip control buttons that combine to provide a more comfortable pipetting experience.
  • QuickStand™ prevents pipette contamination
  • Variable speed control
  • Cordless and rechargable
  • Provides 8 hours of continuous operation
  • Fits serological pipettes from 1 to 100 ml
  • Ambidexterous
Retractable Quickstand™ allows the instrument to be set down on the bench without the risk of contaminating pipettes.
Specifications
  • Description:
    Charger