Order Entry
Mexico
Orders LinkContactUsLinkComponent
LumiLoupe Plus
LumiLoupe Plus
Catalog # 470227-872
Supplier:   CARSON OPTICAL
 :  LO-10
LumiLoupe Plus
Catalog # 470227-872
Supplier:  CARSON OPTICAL
LumiLoupe Plus
LumiLoupe Plus
LumiLoupe Plus
Price
$282.51
Log in for availability
Each (1UN)
Product Details & Documents
A loupe with a transparent base to allow ambient light to illuminate the viewing object.
  • Allows for viewing of an object's fine detail
  • Dual lenses for close-up viewing and less distortion
Specifications
  • Magnification:
    11.5×
  • Description:
    LumiLoupe Plus