Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

VWR® Spinbar® Micro Stir Bars

Stir Bar, 8 mm (⁵⁄₁₆"), 3 mm (1/8"), PTFE
Catalog # 76001-878
Supplier:   VWR International
VWR® Spinbar® Micro Stir Bars
Catalog # 76001-878
Supplier:  VWR International
VWR® Spinbar®  Micro Stir Bars
VWR® Spinbar® Micro Stir Bars

Stir Bar, 8 mm (⁵⁄₁₆"), 3 mm (1/8"), PTFE

Price
$42.75
Log in for availability
Each (1UN) Case de 50 (1UN)
Product Details & Documents

Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes, and gradient makers.

  • Particularly useful for environmental testing
  • Valuable for preparing and evaluating small sample volumes
  • Teflon® PTFE encapsulated
  • Autoclavable

Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.

Specifications
  • Coating:
    PTFE
  • Ø:
    3 mm (1/8")
  • Description:
    Stir Bar
  • Length:
    8 mm (⁵⁄₁₆")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.