Objectives for REVOLVE Microscopes, Echo
Fluorite phase objective, 17 mm, 0.13 NA
75992-490
Supplier #: JMAG0043
Documents
Echo Revolve
Product Literature
- Magnification:4×
- Description:Fluorite phase objective
- Aperture:0.13 NA
- Working Distance:17 mm