Objectives for REVOLVE Microscopes, Echo
Fluorite ELWD phase objective, 3.0 - 4.2 mm, 0.60 NA
Catalog # 75990-662
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0404
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:40×
- Description:Fluorite ELWD phase objective
- Aperture:0.60 NA
- Working Distance:3.0 - 4.2 mm