NeutraKlean Detergent, Contec®
Product Details & Documents
Contec® NeutraKlean is a gentle and low-foaming detergent designed for the cleaning of life science cleanrooms.
- Gentle and low-foaming detergent is ideal for low level cleaning in life science cleanrooms
- Neutral pH makes it suitable for use in all areas of cleanroom
- Filtered to 0.2 microns in a ISO Class 7 cleanroom ensures the product is free from contamination and particulates
- Irradiated at no less than 25 kGy and provided with lot specific sterility certification
- Low level of surfactant provides low residue which can be easily removed
- Trigger spray and "bag in bottle" protected system prevents bottle contents from becoming contaminated during use
With a pH after irradiation between 6.5 and 7.5, NeutraKlean is ideal for carrying out the low-level cleaning required after maintenance, and to remove disinfectant residues and general soil. The use of a surfactant/detergent helps to break the binding of particles and dirt to a surface and allows them to be more easily removed.
Ready to use, the product is available in a 1 L trigger spray with a “bag in bottle” system to protect the contents during use. Both the sterile and filtered products are provided double bagged to aid the transfer of the product into a pharmaceutical cleanroom, isolators, and RABS. The linear tear bags are easy to open, even when wearing gloves.
Specifications
- Description:Irradiated NeutraKlean solution, neutral detergent with purified water
- Size:5 L
- Packaging:Bottle, capped