Order Entry
Mexico
Orders LinkContactUsLinkComponent
Extracta DBS, Quantabio

Extracta DBS, Quantabio

Extracta DBS PCR-ready genomic DNA from dried blood spots, 10 ml
Catalog # 75877-508
Supplier #:  95171-010
Extracta DBS, Quantabio
Catalog # 75877-508
CAS Number:  
Extracta DBS, Quantabio
Extracta DBS, Quantabio

Extracta DBS PCR-ready genomic DNA from dried blood spots, 10 ml

CAS Number:  
Price
$1,621.41
Log in for availability
Each (10ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Extracta DBS is a rapid and easy to use extractions reagent optimized for the extraction of DNA from dried blood spot punches

  • Optimized for DNA extraction from dried blood spot punches
  • Single reagent for PCR-ready DNA in less than 30 minutes
  • Maximized assay sensitivity, lower Ct’s, when combined with Quantabio ToughMix
  • Compatible with high-throughput automation for PCR, qPCR and NGS applications
  • Reliable, Consistent Lot-to-Lot Performance

Extracta DBS is a ready-to-use DNA extraction reagent for rapid and efficient recovery of PCR-ready DNA from dried blood spots (DBS) on Guthrie cards or Whatman 903 filter paper. This patented single-solution process produces DNA eluates that are substantially free of PCR inhibitors and compatible with a variety of end-point PCR, real-time PCR and Next Generation Sequencing (NGS) or Sanger Sequencing (1-3) reagents. Application of Extracta DBS with PerfeCta® qPCR ToughMix® or PerfeCta® MultiPlex qPCR ToughMix enables accurate and reproducible quantification of DNA sequences in blood using TaqMan® hydrolysis probe real-time qPCR.

The resulting extract is immediately usable for downstream applications such as real-time PCR.

Specifications

Specifications

  • Description:
    Extracta DBS PCR-ready genomic DNA from dried blood spots
  • Volume:
    10 ml
  • Cat. No.:
    75877-508
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Extraction for direct PCR
  • Description:
    Extracta DBS PCR-ready Genomic DNA
  • Sample type:
    Blood (dried) on filter cards

Specifications