Order Entry
Mexico
Orders LinkContactUsLinkComponent
Extracta DBS, Quantabio

Extracta DBS, Quantabio

Extracta DBS PCR-ready genomic DNA from dried blood spots, 500 ml
Catalog # 75877-506
Supplier #:  95171-500
Extracta DBS, Quantabio
Catalog # 75877-506
CAS Number:  
Extracta DBS, Quantabio
Extracta DBS, Quantabio

Extracta DBS PCR-ready genomic DNA from dried blood spots, 500 ml

CAS Number:  
Price
$20,852.04
Log in for availability
Each (500ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Extracta DBS is a rapid and easy to use extractions reagent optimized for the extraction of DNA from dried blood spot punches

  • Optimized for DNA extraction from dried blood spot punches
  • Single reagent for PCR-ready DNA in less than 30 minutes
  • Maximized assay sensitivity, lower Ct’s, when combined with Quantabio ToughMix
  • Compatible with high-throughput automation for PCR, qPCR and NGS applications
  • Reliable, Consistent Lot-to-Lot Performance

Extracta DBS is a ready-to-use DNA extraction reagent for rapid and efficient recovery of PCR-ready DNA from dried blood spots (DBS) on Guthrie cards or Whatman 903 filter paper. This patented single-solution process produces DNA eluates that are substantially free of PCR inhibitors and compatible with a variety of end-point PCR, real-time PCR and Next Generation Sequencing (NGS) or Sanger Sequencing (1-3) reagents. Application of Extracta DBS with PerfeCta® qPCR ToughMix® or PerfeCta® MultiPlex qPCR ToughMix enables accurate and reproducible quantification of DNA sequences in blood using TaqMan® hydrolysis probe real-time qPCR.

The resulting extract is immediately usable for downstream applications such as real-time PCR.

Specifications

Specifications

  • Description:
    Extracta DBS PCR-ready genomic DNA from dried blood spots
  • Volume:
    500 ml
  • Cat. No.:
    75877-506
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Extraction for direct PCR
  • Description:
    Extracta DBS PCR-ready Genomic DNA
  • Sample type:
    Blood (dried) on filter cards

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.