Iron Meteorite
Iron Meteorite, ⅜ - ¾"
Catalog # 470225-098
Supplier: Avantor
Jump to:
Product Details & Documents
Fragments in natural condition, many showing concave forms and fusion crusts. Sizes are approximate.
- Available in two sizes
- Slicked in oil to prevent rusting
- Protected in bubble wrap and a sealed bag
Specifications
- Description:Iron Meteorite
- Specimen Dimensions:⅜ - ¾"