Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Polyethylene Transfer Pipettes

1.5 ml Small Bulb Extended Tip, 104 mm, 50, Non-Graduated , 1.0 ml
  470225-068
Polyethylene Transfer Pipettes
  470225-068
Polyethylene Transfer Pipettes
Polyethylene Transfer Pipettes

1.5 ml Small Bulb Extended Tip, 104 mm, 50, Non-Graduated , 1.0 ml

 
$1,667.30
   
Case (500UN)
 

Disposable transfer pipettes for liquid handling.

  • Transparent, non-toxic, and inert
  • Precise, uniform drop size
  • Shatter-proof and unbreakable
  • Can be used with liquid nitrogen
  • Heat sealable for liquid storage

Unbreakable, one-piece pipettes eliminate the need for rubber bulbs, broken glass, and exposure to infected material.

Ideal for microbiology, blood banking, hematology, immunology, and urinalysis. Pipettes can also be heat sealed for airtight sample storage.

 : Available packaging includes nonsterile bulk, sterile individually wrapped, or sterile packs of 20.

 
  • Description:
    1.5 ml Small Bulb Extended Tip
  • Overall Length:
    104 mm
  • Graduations:
    Non-Graduated
  • Bulb Draw:
    1.0 ml
  • Drops per mL:
    50