Order Entry
Mexico
Orders LinkContactUsLinkComponent
Canonical URL fetch failed:ERROR

MacroTubes, 5 ml

Lock Clip for Snap Cap Tubes, —, 20 per Pack
Catalog # 470225-024
Supplier:   MTC BIO, INC.
Supplier #:  C2586_WARDS
MacroTubes, 5 ml
Catalog # 470225-024
Supplier:  MTC BIO, INC.
MacroTubes, 5 ml
MacroTubes, 5 ml

Lock Clip for Snap Cap Tubes, —, 20 per Pack

Price
$821.90
Log in for availability
Paquete de 20
Product Details & Documents

The perfect tubes for intermediate volumes.

  • Uniform wall thickness
  • Appropriate for storage from –86 to 100 °C
  • Color-code samples for easy identification
  • RNAse-, DNAse-, Pyrogen-, and Cytotoxin-free
  • Snap-cap or screw-cap available

Made of maximum clarity, low-binding virgin polypropylene, these tubes are sized to be compatible with all standard equipment.

They're a valuable resource for boiling and for sample preservation.

Specifications
  • Description:
    Lock Clip for Snap Cap Tubes
  • Dimensions:
  • Packaging:
    20 per Pack
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.