Order Entry
Mexico
Orders LinkContactUsLinkComponent
SANI-STICK Sampling Kits, Labplas
SANI-STICK kit with wet sponge on handle with DE-neutralizing buffer, 600 ml, 140×279 mm (5.5×11")
Catalog # 75849-340
Supplier:   LABPLAS INC CA
 :  KSS-67310-DE-VW
SANI-STICK Sampling Kits, Labplas
Catalog # 75849-340
Supplier:  LABPLAS INC CA
SANI-STICK Sampling Kits, Labplas
SANI-STICK Sampling Kits, Labplas
SANI-STICK kit with wet sponge on handle with DE-neutralizing buffer, 600 ml, 140×279 mm (5.5×11")
Price
$9,318.48
Log in for availability
Case (100UN)
Product Details & Documents
The SANI-STICK is designed to collect samples to detect the presence of microbiological contaminations on any surface.
  • Allows swabbing of hard-to-reach areas
  • Convenient and east to use
  • Helps eliminate several time -consuming steps
These sponges on sticks are widely used in the food industry applications such as quality assurance procedures (QA/QC).
Specifications
  • Capacity:
    600 ml
  • Description:
    SANI-STICK kit with wet sponge on handle with DE-neutralizing buffer
  • W×L:
    140×279 mm (5.5×11")