SANI-STICK Sampling Kits, Labplas
SANI-STICK kit with wet sponge on handle with letheen broth, 600 ml, 140×279 mm (5.5×11")
Catalog # 75849-320
Supplier: LABPLAS INC CA
: KSS-67310-LT
Jump to:
Product Details & Documents
The SANI-STICK is designed to collect samples to detect the presence of microbiological contaminations on any surface.
- Allows swabbing of hard-to-reach areas
- Convenient and east to use
- Helps eliminate several time -consuming steps
These sponges on sticks are widely used in the food industry applications such as quality assurance procedures (QA/QC).
Specifications
- Capacity:600 ml
- Description:SANI-STICK kit with wet sponge on handle with letheen broth
- W×L:140×279 mm (5.5×11")



