Repeater® E3 and E3x Pipettors, Eppendorf®
Product Details & Documents
The Repeater® E3/E3x Pipettors, in combination with the Combitips Advanced® pipette tips, creates a versatile dispensing system that functions according to the positive displacement principle.
- Automatic Combitip® recognition
- Ergonomic one-button tip ejector
- Precisely programmable aspiration and dispensing speeds
- Repetitive dispensing modes
- Wide dispensing range: 1 μl to 50 ml, in 100-nanoliter increments
- Safe and ergonomic operation with 9 different selectable languages
These devices provide precise and accurate dispensing of viscous solutions (e.g. glycerol) or liquids with high vapor pressure (e.g. ethanol). Contamination-free dispensing for toxic, radioactive, and other hazardous liquids (all available modes). Large illuminated color display reduces eye strain when reading. Motor-driven function reduces human error to a minimum, and reduces the risk of repetitive strain injuries (RSI).
It is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results.
Documents
Specifications
- Description:Repeater® E3 Bundle, Including Charger Stand
Accessories
Product Information
- Catalog #AvailabilityPrice
- Charger stand 2 for one Repeater® E3/E3x