Order Entry
Mexico
Orders LinkContactUsLinkComponent
Density Sets
Density Sets
Density Set, Brass
Catalog # 470221-112
Número de proveedor #:  PH0113J
Density Sets
Catalog # 470221-112
Density Sets
Density Sets
Density Set, Brass
Price
$2,525.92
Log in for availability
Each (1UN)
Product Details & Documents
Students Learn That the Density of a Substance is Constant, Even as Its Mass and Volume Change.
  • Each set has 12 cylindrical bars
  • Bars are same diameter but different lengths
  • Each set comes in an attractive wooden holder
Specifications
  • Description:
    Density Set, Brass